4-Diethylcarbamoyl-2-methoxybenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Diethylcarbamoyl)-2-methoxyphenylboronic acid

13476-1G 13476-25G 13476-500MG 13476-5G
76907-180EA 201.58 USD
76907-180 76907-182 76907-184 76907-186
4-Diethylcarbamoyl-2-methoxybenzeneboronic acid ≥97%
4-Diethylcarbamoyl-2-methoxybenzeneboronic acid
Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027199
CAS Number: 913835-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR