3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine, 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 3-(Pyrrolidinomethyl)phenylboronic acid, pinacol ester

13364-1G 13364-25G 13364-5G
76907-160EA 64.72 USD
76907-160 76907-162 76907-164
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester ≥95%
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₂
MW: 287.205 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08741425
CAS Number: 884507-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR