3-Carboxy-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Borono-2-methoxybenzoic acid, 3-Carboxy-4-methoxybenzeneboronic acid

13339-25G 13339-5G
76907-148EA 209 USD
76907-148 76907-150
3-Carboxy-4-methoxyphenylboronic acid ≥97%
3-Carboxy-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MW: 195.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203322
CAS Number: 913836-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR