3-Bromo-2-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromopyridine-2-carboxaldehyde

13298-25G 13298-5G
76907-138EA 488.02 USD
76907-138 76907-140
3-Bromo-2-formylpyridine ≥97%
3-Bromo-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781220
CAS Number: 405174-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR