4-Amino-3-fluorophenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester
13167-25G
13167-5G
76907-118EA
375.56
USD
76907-118
76907-120
4-Amino-3-fluorophenylboronic acid pinacol ester ≥97%
4-Amino-3-fluorophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09033884 CAS Number: 819058-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...