4-Amino-3-chlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Amino-3-chlorobenzeneboronic acid, pinacol ester, 4-Amino-3-chlorophenylboronic Acid Pinacol Ester

13150-1G 13150-25G 13150-5G
76907-108EA 62.6 USD
76907-108 76907-110 76907-112
4-Amino-3-chlorophenylboronic acid pinacol ester ≥97%
4-Amino-3-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD09998359
CAS Number: 721960-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR