3-Iodo-2,5-dimethylthiophene ≥97%
Supplier: AOBChem USA
|
Warning
|
131119-1G
131119-25G
131119-500MG
131119-5G
76907-092EA
257.8
USD
76907-092
76907-094
76907-096
76907-098
3-Iodo-2,5-dimethylthiophene ≥97%
3-Iodo-2,5-dimethylthiophene
Precaución: for Research Use Only
Formula:
C₆H₇IS MW: 238.089 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22369798 CAS Number: 40197-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...