3,5-Dibromo-2-fluoro-6-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

13077-25G 13077-5G
76907-084EA 453.01 USD
76907-084 76907-086
3,5-Dibromo-2-fluoro-6-methylpyridine ≥97%
3,5-Dibromo-2-fluoro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄NBr₂F
Storage Temperature: Ambient
MDL Number: MFCD09475419
CAS Number: 632628-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR