4-Chloro-3,5-dimethylphenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

12777-1G 12777-25G 12777-5G
76907-050EA 62.6 USD
76907-050 76907-052 76907-054
4-Chloro-3,5-dimethylphenylboronic acid ≥95% research grade
4-Chloro-3,5-dimethylphenylboronic acid
Formula: C₈H₁₀BClO₂
MW: 184.4278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566596
CAS Number: 1056475-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR