4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Acetoxyphenylboronic acid pinacol ester, 2-(4-Acetoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl Acetate, 4-Acetoxyphenylboronic acid, pinacol ester, 4-Acetoxyphenylboronic acid Pinacol Ester

12668-25G 12668-5G
76907-036EA 488.02 USD
76907-036 76907-038
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD03453060
CAS Number: 480424-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR