4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Tert-butylcarbonylamino)phenylboronic acid, 4-Pivalamidobenzeneboronic acid

12559-1G 12559-25G 12559-500MG 12559-5G
76907-020EA 266.29 USD
76907-020 76907-022 76907-024 76907-026
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid ≥97%
4-[(2,2-Dimethylpropanoyl)amino]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235071
CAS Number: 182344-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR