4-(Propionyl)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Propionyl)phenyboronic acid, (4-Propionylphenyl)boronic acid

12533-1G 12533-25G 12533-5G
76907-012EA 71.09 USD
76907-012 76907-014 76907-016
4-(Propionyl)benzeneboronic acid ≥97%
4-(Propionyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04039038
CAS Number: 186498-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR