3-(Methylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid

12473-25G 12473-5G
76906-996EA 346.92 USD
76906-996 76906-998
3-(Methylcarbamoyl)phenylboronic acid ≥97%
3-(Methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04038918
CAS Number: 832695-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR