3-(Isobutylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Isobutylaminocarbonyl)phenylboronic acid

12389-25G 12389-5G
76906-976EA 310.85 USD
76906-976 76906-978
3-(Isobutylcarbamoyl)phenylboronic acid ≥97%
3-(Isobutylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD04115692
CAS Number: 723282-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR