3-Hydroxy-4-methylbenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

12313-1G 12313-25G 12313-500MG 12313-5G
76906-964EA 171.87 USD
76906-964 76906-966 76906-968 76906-970
3-Hydroxy-4-methylbenzeneboronic acid ≥97%
3-Hydroxy-4-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD01074652
CAS Number: 216019-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR