3-(Methoxymethyl)phenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxymethylphenylboronic acid, (3-(Methoxymethyl)phenyl)boronic acid

12162-1G 12162-25G 12162-5G
76906-926EA 73.21 USD
76906-926 76906-928 76906-930
3-(Methoxymethyl)phenylboronic acid ≥95% research grade
3-(Methoxymethyl)phenylboronic acid
Formula: C₈H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03412069
CAS Number: 142273-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR