4-(Isobutylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Isobutylcarbamoyl)phenyl)boronic acid, 4-(Isobutylaminocarbonyl)phenylboronic acid, 4-(Isobutylcarbamoyl)benzeneboronic acid

12120-25G 12120-5G
76906-906EA 587.74 USD
76906-906 76906-908
4-(Isobutylcarbamoyl)phenylboronic acid ≥97%
4-(Isobutylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411955
CAS Number: 850568-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR