3'-Hydroxy-4'-methoxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-4-methoxyacetophenone

12090-1G 12090-25G 12090-500MG 12090-5G
76906-894EA 185.66 USD
76906-894 76906-896 76906-898 76906-900
3'-Hydroxy-4'-methoxyacetophenone ≥97%
3'-Hydroxy-4'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00182975
CAS Number: 6100-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR