3-Fluoro-4-isopropoxyphenylboronicacid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-4-isopropoxyphenylboronic acid

12062-25G 12062-5G
76906-886EA 375.56 USD
76906-886 76906-888
3-Fluoro-4-isopropoxyphenylboronicacid ≥97%
3-Fluoro-4-isopropoxyphenylboronicacid
Formula: C₉H₁₂BFO₃
MW: 198.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865163
CAS Number: 480438-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR