3-(Ethoxycarbonyl)-5-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11928-25G 11928-5G
76906-858EA 198.39 USD
76906-858 76906-860
3-(Ethoxycarbonyl)-5-nitrophenylboronic acid ≥97%
3-(Ethoxycarbonyl)-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₆
Storage Temperature: Ambient
MDL Number: MFCD03788422
CAS Number: 850568-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR