3,4-Dichlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

11891-25G 11891-5G
76906-844EA 346.92 USD
76906-844 76906-846
3,4-Dichlorophenylboronic acid pinacol ester ≥97%
3,4-Dichlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05663853
CAS Number: 401797-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR