3-Fluoro-5-hydroxypyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-5-pyridinol, 5-Fluoropyridin-3-ol

11587-25G 11587-5G
76906-784EA 198.39 USD
76906-784 76906-786
3-Fluoro-5-hydroxypyridine ≥95%
3-Fluoro-5-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
Storage Temperature: Ambient
MDL Number: MFCD03095199
CAS Number: 209328-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR