3-Chloro-4-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11467-25G 11467-5G
76906-774EA 268.41 USD
76906-774 76906-776
3-Chloro-4-isopropoxyphenylboronic acid ≥97%
3-Chloro-4-isopropoxyphenylboronic acid
Formula: C₉H₁₂BClO₃
MW: 214.4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974065
CAS Number: 480438-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR