4-(Dipropylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Dipropylcarbamoyl)phenyl)boronic acid, 4-(N,N-Dipropylaminocarbonyl)phenylboronic acid, 4-(N,N-Dipropylaminocarbonyl)benzeneboronic acid

11435-25G 11435-5G
76906-740EA 630.18 USD
76906-740 76906-742
4-(Dipropylcarbamoyl)phenylboronic acid ≥97%
4-(Dipropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MW: 249.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659883
CAS Number: 850568-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR