4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

114213-1G 114213-25G 114213-500MG 114213-5G
76906-728EA 132.62 USD
76906-728 76906-730 76906-732 76906-734
4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid ≥97%
4-[(3-Fluorobenzyl)oxy]benzenecarboxylic acid
Formula: C₁₄H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD05663826
CAS Number: 405-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR