3-Iodo-2-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

11378-1G 11378-25G 11378-5G
76906-720EA 94.43 USD
76906-720 76906-722 76906-724
3-Iodo-2-methylpyridine ≥95%
3-Iodo-2-methylpyridine
Formula: C₆H₆IN
MW: 219.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09702485
CAS Number: 15112-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR