3-Amino-4-chlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Amino-4-chlorobenzeneboronic acid pinacol ester, 3-Amino-4-chlorophenylboronic acid, pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

11361-25G 11361-5G
76906-712EA 502.87 USD
76906-712 76906-714
3-Amino-4-chlorophenylboronic acid pinacol ester ≥97%
3-Amino-4-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD06659926
CAS Number: 850567-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR