4-(Cyclopropylaminocarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(Cyclopropylcarbamoyl)benzeneboronic acid, 4-(Cyclopropylaminocarbonyl)phenylboronic acid

11287-25G 11287-5G
76906-704EA 219.61 USD
76906-704 76906-706
4-(Cyclopropylaminocarbonyl)phenylboronic acid ≥97% research grade
4-(Cyclopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03788419
CAS Number: 515140-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR