3-Carboxy-5-nitrophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Carboxy-5-nitrobenzeneboronic acid pinacol ester, 3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-nitrophenylboronic acid, pinacol ester

11273-25G 11273-5G
76906-694EA 453.01 USD
76906-694 76906-696
3-Carboxy-5-nitrophenylboronic acid pinacol ester ≥97%
3-Carboxy-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₆
MW: 293.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411932
CAS Number: 377780-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR