3-Bromo-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Bromopyridine-4-boronic acid pinacol ester

11231-25G 11231-5G
76906-676EA 531.52 USD
76906-676 76906-678
3-Bromo-4-pyridineboronic acid pinacol ester ≥97%
3-Bromo-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BBrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06798250
CAS Number: 458532-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR