1-(Benzyloxy)-2-bromo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzoxyl-3-bromotoluene

11207-25G 11207-5G
76906-662EA 346.92 USD
76906-662 76906-664
1-(Benzyloxy)-2-bromo-4-methylbenzene ≥97%
1-(Benzyloxy)-2-bromo-4-methylbenzene
Formula: C₁₄H₁₃BrO
MW: 277.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753753
CAS Number: 2830-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR