4-Bromo-2,3-dichlorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

111365-1G 111365-25G 111365-5G
76906-642EA 109.28 USD
76906-642 76906-644 76906-646
4-Bromo-2,3-dichlorobenzonitrile ≥97%
4-Bromo-2,3-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrCl₂N
Storage Temperature: Ambient
CAS Number: 1160574-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR