3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-(Bromomethyl)phenyl)-5,5-dimethyl-1,3,2-dioxaborinane

11087-25G 11087-5G
76906-616EA 396.78 USD
76906-616 76906-618
3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester ≥97%
3-(Bromomethyl)benzeneboronic acid, neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BBrO₂
MW: 282.969 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04973735
CAS Number: 223799-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR