4-Benzyloxy-3,5-dichlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11079-1G 11079-25G 11079-5G
76906-604EA 106.09 USD
76906-604 76906-606 76906-608
4-Benzyloxy-3,5-dichlorophenylboronic acid ≥97%
4-Benzyloxy-3,5-dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04039002
CAS Number: 1003298-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR