5-Bromo-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Bromopyridine-5-boronic acid pinacol ester

11065-25G 11065-5G
76906-596EA 289.63 USD
76906-596 76906-598
5-Bromo-3-pyridineboronic acid pinacol ester ≥97%
5-Bromo-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BBrNO₂
Storage Temperature: Ambient
MDL Number: MFCD06798248
CAS Number: 452972-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR