4-Benzyloxy-2,6-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Benzyloxy)-2,6-difluorophenyl)boronic acid

11034-1G 11034-25G 11034-500MG 11034-5G
76906-584EA 124.13 USD
76906-584 76906-586 76906-588 76906-590
4-Benzyloxy-2,6-difluorophenylboronic acid ≥97%
4-Benzyloxy-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MW: 264.0325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026734
CAS Number: 156635-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR