4-Bromo-2,3-difluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

11003-25G 11003-5G
76906-576EA 418 USD
76906-576 76906-578
4-Bromo-2,3-difluorobenzeneboronic acid ≥97%
4-Bromo-2,3-difluorobenzeneboronic acid
Formula: C₆H₄BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02093065
CAS Number: 374790-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR