4-(tert-Butylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(tert-Butylcarbamoyl)benzeneboronic acid, (4-(tert-Butylcarbamoyl)phenyl)boronic acid, 4-(tert-Butylaminocarbonyl)phenylboronic acid

10979-25G 10979-5G
76906-562EA 310.85 USD
76906-562 76906-564
4-(tert-Butylcarbamoyl)phenylboronic acid ≥97%
4-(tert-Butylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788413
CAS Number: 850568-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR