3-(Benzylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N-Benzylaminocarbonyl)phenylboronic acid

10977-25G 10977-5G
76906-554EA 488.02 USD
76906-554 76906-556
3-(Benzylcarbamoyl)phenylboronic acid ≥97%
3-(Benzylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04115688
CAS Number: 625470-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR