(3-Acetamidophenyl)boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 3-Acetamidophenylboronic acid Pinacol ester, 3-Acetamidophenylboronic acid, pinacol estet, 2-(3-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide

10742-25G 10742-5G
76906-476EA 275.84 USD
76906-476 76906-478
(3-Acetamidophenyl)boronic acid pinacol ester ≥97%
(3-Acetamidophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MW: 261.1245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789261
CAS Number: 480424-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR