3-(tert-Butyldimethylsilyloxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-(tert-Butyl dimethylsiloxy)phenyl boronic acid, (3-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid, 3-(tert-Butyldimethylsiloxy)benzeneboronic acid

10454-25G 10454-5G
76906-400EA 247.19 USD
76906-400 76906-402
3-(tert-Butyldimethylsilyloxy)phenylboronic acid ≥97% research grade
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
Formula: C₁₂H₂₁BO₃Si
Storage Temperature: Ambient
MDL Number: MFCD03701505
CAS Number: 261621-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR