3-Methoxy-4-carboxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

10281-25G 10281-5G
76906-344EA 403.15 USD
76906-344 76906-346
3-Methoxy-4-carboxyphenylboronic acid ≥97% research grade
3-Methoxy-4-carboxyphenylboronic acid
Formula: C₈H₉BO₅
Storage Temperature: Ambient
MDL Number: MFCD06656278
CAS Number: 851335-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR