3-Methoxy-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxy-4-methylbenzeneboronic acid

10278-25G 10278-5G
76906-336EA 233.4 USD
76906-336 76906-338
3-Methoxy-4-methylphenylboronic acid ≥97%
3-Methoxy-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD08274479
CAS Number: 917757-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR