4-(Piperidine-1-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-[(Piperidin-1-yl)carbonyl]benzeneboronic acid

10256-25G 10256-5G
76906-326EA 325.7 USD
76906-326 76906-328
4-(Piperidine-1-carbonyl)phenylboronic acid ≥97%
4-(Piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₆BNO₃
MW: 233.0713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411951
CAS Number: 389621-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR