4-Bromo-2-chloro-5-(trifluoromethyl)aniline ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-chloro-5-(trifluoromethyl)benzenamine

10017-25G 10017-5G
76906-246EA 453.01 USD
76906-246 76906-248
4-Bromo-2-chloro-5-(trifluoromethyl)aniline ≥95%
4-Bromo-2-chloro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃N
Storage Temperature: Ambient
MDL Number: MFCD12547854
CAS Number: 104460-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR