4-Acetyl-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10013-25G 10013-5G
76906-240EA 559.1 USD
76906-240 76906-242
4-Acetyl-3-fluorophenylboronic acid ≥97%
4-Acetyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₃
MW: 181.9600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796422
CAS Number: 481725-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR