2'-Fluoro-4'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

96594-25G 96594-5G
76893-984EA 251.44 USD
76893-984 76893-986
2'-Fluoro-4'-methylacetophenone ≥97%
2'-Fluoro-4'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD03094331
CAS Number: 29427-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR