1-Chloro-2-ethoxy-5-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

94997-1G 94997-25G 94997-500MG 94997-5G
76893-970EA 127.31 USD
76893-970 76893-972 76893-974 76893-976
1-Chloro-2-ethoxy-5-fluoro-3-iodobenzene ≥97%
1-Chloro-2-ethoxy-5-fluoro-3-iodobenzene
Formula: C₈H₇ClFIO
MW: 300.496 g/mol
Storage Temperature: Ambient
CAS Number: 2385991-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR