1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate ≥95%

Supplier: AOBChem USA

Warning

93385-25G 93385-5G 93385-1G
76893-944EA 555.92 USD
76893-944 76893-946 77640-208
1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate ≥95%
1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₂N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD32862308
CAS Number: 1872262-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR