2,4-Dichlorothioanisole ≥97%

Supplier: AOBChem USA

Warning

91868-25G 91868-5G
76893-904EA 484.84 USD
76893-904 76893-906
2,4-Dichlorothioanisole ≥97%
2,4-Dichlorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆Cl₂S
Storage Temperature: Ambient
CAS Number: 65909-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR